Assay Details
| Gene Symbol: Kiaa1715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498321.2 | XP_003498369.1 | 5 - 6 | 418 | 123 | |
| XM_003498322.2 | XP_003498370.1 | 5 - 6 | 418 | 123 | ||
| GenBank mRNA | JI885934.1 | - | 5 - 6 | 355 | 123 | |
| JP055124.1 | - | 5 - 6 | 355 | 123 | ||
More Information
Set Membership: |
|

Kiaa1715
Genomic Map