Assay Details
| Gene Symbol: Kiaa1715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498321.2 | XP_003498369.1 | 7 - 8 | 549 | 98 | |
| XM_003498322.2 | XP_003498370.1 | 7 - 8 | 549 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kiaa1715| Gene Symbol: Kiaa1715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498321.2 | XP_003498369.1 | 7 - 8 | 549 | 98 | |
| XM_003498322.2 | XP_003498370.1 | 7 - 8 | 549 | 98 | ||