Assay Details
| Gene Symbol: Rad51ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503049.1 | XP_003503097.1 | 4 - 5 | 2834 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad51ap2| Gene Symbol: Rad51ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503049.1 | XP_003503097.1 | 4 - 5 | 2834 | 98 | |
Set Membership: |
|