Assay Details
| Gene Symbol: LOC100763924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654679.1 | XP_007652869.1 | 6 - 7 | 739 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763924| Gene Symbol: LOC100763924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654679.1 | XP_007652869.1 | 6 - 7 | 739 | 90 | |