Assay Details
| Gene Symbol: Slc25a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512169.2 | XP_003512217.1 | 2 - 3 | 363 | 61 | |
| GenBank mRNA | JI882783.1 | - | 2 - 3 | 301 | 61 | |
| JP037430.1 | - | 2 - 3 | 301 | 61 | ||
More Information
Set Membership: |
|

Slc25a20
Genomic Map