Assay Details
| Gene Symbol: Cmtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499677.2 | XP_003499725.1 | 4 - 5 | 654 | 73 | |
| XM_007640410.1 | XP_007638600.1 | 3 - 4 | 559 | 73 | ||
| GenBank mRNA | JI865217.1 | - | 1 - 2 | 110 | 73 | |
| JP060466.1 | - | 1 - 2 | 112 | 73 | ||
More Information
Set Membership: |
|

Cmtm3
Genomic Map