Assay Details
| Gene Symbol: Nptxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650060.1 | XP_007648250.1 | 4 - 5 | 701 | 77 | |
| GenBank mRNA | JI868021.1 | - | 1 - 2 | 131 | 77 | |
More Information
Set Membership: |
|

Nptxr
Genomic Map