Assay Details
| Gene Symbol: Cpa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497539.1 | XP_003497587.1 | 5 - 6 | 488 | 60 | |
| XM_003497540.1 | XP_003497588.1 | 4 - 5 | 389 | 60 | ||
More Information
Set Membership: |
|

Cpa4
Genomic Map