Assay Details
| Gene Symbol: Abcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506537.2 | XP_003506585.1 | 7 - 8 | 2155 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcd1| Gene Symbol: Abcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506537.2 | XP_003506585.1 | 7 - 8 | 2155 | 59 | |
Set Membership: |
|