Assay Details
| Gene Symbol: Rnaseh2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512924.2 | XP_003512972.1 | 2 - 3 | 450 | 87 | |
| XM_007653318.1 | XP_007651508.1 | 2 - 3 | 451 | 87 | ||
| GenBank mRNA | JI876168.1 | - | 2 - 3 | 246 | 87 | |
| JP065098.1 | - | 2 - 3 | 246 | 87 | ||
More Information
Set Membership: |
|

Rnaseh2c
Genomic Map