Assay Details
| Gene Symbol: Cxcl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007608718.1 | XP_007606908.1 | 2 - 3 | 312 | 64 | |
| GenBank mRNA | JI888463.1 | - | 2 - 3 | 312 | 64 | |
More Information
Set Membership: |
|

Cxcl16
Genomic Map