Assay Details
| Gene Symbol: Eif4ebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640645.1 | XP_007638835.1 | 2 - 3 | 264 | 73 | |
| GenBank mRNA | JI894830.1 | - | 1 - 2 | 196 | 73 | |
More Information
Set Membership: |
|

Eif4ebp2
Genomic Map