Assay Details
Gene Symbol: Rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504641.2 | XP_003504689.1 | 3 - 4 | 518 | 60 | |
GenBank mRNA | JI879766.1 | - | 3 - 4 | 506 | 60 | |
JP058501.1 | - | 3 - 4 | 506 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
