Assay Details
| Gene Symbol: Aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509931.2 | XP_003509979.1 | 5 - 6 | 927 | 68 | |
| GenBank mRNA | JI880559.1 | - | 5 - 6 | 900 | 68 | |
More Information
Set Membership: |
|

Aip
Genomic Map