Assay Details
| Gene Symbol: Nab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507486.2 | XP_003507534.1 | 4 - 5 | 1311 | 62 | |
| XM_003507487.2 | XP_003507535.1 | 4 - 5 | 1311 | 62 | ||
| GenBank mRNA | JI870425.1 | - | 4 - 5 | 1141 | 62 | |
| JP046212.1 | - | 3 - 4 | 242 | 62 | ||
More Information
Set Membership: |
|

Nab2
Genomic Map