Assay Details
| Gene Symbol: Setbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645379.1 | XP_007643569.1 | 5 - 6 | 3405 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Setbp1| Gene Symbol: Setbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645379.1 | XP_007643569.1 | 5 - 6 | 3405 | 79 | |
Set Membership: |
|