Assay Details
| Gene Symbol: Gabrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641080.1 | XP_007639270.1 | 6 - 7 | 656 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrr1| Gene Symbol: Gabrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641080.1 | XP_007639270.1 | 6 - 7 | 656 | 78 | |
Set Membership: |
|