Assay Details
| Gene Symbol: Camlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498145.2 | XP_003498193.1 | 3 - 4 | 727 | 71 | |
| XM_007623123.1 | XP_007621313.1 | 3 - 4 | 683 | 71 | ||
| GenBank mRNA | JI884483.1 | - | 3 - 4 | 566 | 71 | |
More Information
Set Membership: |
|

Camlg
Genomic Map