Assay Details
| Gene Symbol: Ndufa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508802.2 | XP_003508850.1 | 1 - 2 | 169 | 76 | |
| GenBank mRNA | JI880509.1 | - | 1 - 2 | 175 | 76 | |
| JP050448.1 | - | 1 - 2 | 175 | 76 | ||
More Information
Set Membership: |
|

Ndufa6
Genomic Map