Assay Details
| Gene Symbol: Gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640786.1 | XP_007638976.1 | 3 - 4 | 331 | 78 | |
| XM_007640787.1 | XP_007638977.1 | 3 - 4 | 335 | 78 | ||
| GenBank mRNA | JP060605.1 | - | 3 - 4 | 228 | 78 | |
More Information
Set Membership: |
|

Gmfb
Genomic Map