Assay Details
| Gene Symbol: Gas2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507334.2 | XP_003507382.1 | 8 - 9 | 892 | 75 | |
| XM_007647898.1 | XP_007646088.1 | 9 - 10 | 952 | 75 | ||
| XM_007647899.1 | XP_007646089.1 | 10 - 11 | 1203 | 75 | ||
| XM_007647900.1 | XP_007646090.1 | 9 - 10 | 931 | 75 | ||
| GenBank mRNA | JI878299.1 | - | 8 - 9 | 833 | 75 | |
| JP055906.1 | - | 8 - 9 | 833 | 75 | ||
More Information
Set Membership: |
|

Gas2l3
Genomic Map