Assay Details
| Gene Symbol: Fn3krp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509988.2 | XP_003510036.1 | 5 - 6 | 637 | 83 | |
| GenBank mRNA | JI862400.1 | - | 2 - 3 | 211 | 83 | |
| JP048629.1 | - | 2 - 3 | 211 | 83 | ||
More Information
Set Membership: |
|

Fn3krp
Genomic Map