Assay Details
| Gene Symbol: LOC100764018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649406.1 | XP_007647596.1 | 1 - 2 | 122 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100764018| Gene Symbol: LOC100764018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649406.1 | XP_007647596.1 | 1 - 2 | 122 | 87 | |
Set Membership: |
|