Assay Details
| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505867.2 | XP_003505915.1 | 2 - 3 | 648 | 93 | |
| XM_007646353.1 | XP_007644543.1 | 2 - 3 | 648 | 93 | ||
| GenBank mRNA | JI875392.1 | - | 1 - 2 | 81 | 93 | |
More Information
Set Membership: |
|

Tcf19
Genomic Map