Assay Details
| Gene Symbol: S100pbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500629.2 | XP_003500677.1 | 2 - 3 | 1103 | 71 | |
| GenBank mRNA | JI895168.1 | - | 1 - 2 | 136 | 71 | |
| JP049900.1 | - | 1 - 2 | 136 | 71 | ||
More Information
Set Membership: |
|

S100pbp
Genomic Map