Assay Details
| Gene Symbol: Coro1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503529.2 | XP_003503577.1 | 6 - 7 | 770 | 58 | |
| XM_007644138.1 | XP_007642328.1 | 6 - 7 | 804 | 58 | ||
| XM_007644139.1 | XP_007642329.1 | 6 - 7 | 784 | 58 | ||
| GenBank mRNA | JI880461.1 | - | 5 - 6 | 854 | 58 | |
| JP037991.1 | - | 5 - 6 | 854 | 58 | ||
More Information
Set Membership: |
|

Coro1c
Genomic Map