Assay Details
| Gene Symbol: Ldoc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515297.2 | XP_003515345.1 | 1 - 2 | 1234 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldoc1l| Gene Symbol: Ldoc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515297.2 | XP_003515345.1 | 1 - 2 | 1234 | 67 | |
Set Membership: |
|