Assay Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512277.2 | XP_003512325.2 | 11 - 12 | 1963 | 67 | |
| GenBank mRNA | JI872395.1 | - | 11 - 12 | 897 | 67 | |
More Information
Set Membership: |
|

Fam50a
Genomic Map