Assay Details
| Gene Symbol: Gfy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508645.1 | XP_003508693.1 | 2 - 3 | 1334 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfy| Gene Symbol: Gfy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508645.1 | XP_003508693.1 | 2 - 3 | 1334 | 105 | |
Set Membership: |
|