Assay Details
| Gene Symbol: Chrac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510999.2 | XP_003511047.1 | 1 - 2 | 193 | 83 | |
| GenBank mRNA | JI889730.1 | - | 1 - 2 | 168 | 83 | |
| JP044942.1 | - | 1 - 2 | 168 | 83 | ||
More Information
Set Membership: |
|

Chrac1
Genomic Map