Assay Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647804.1 | XP_007645994.1 | 7 - 8 | 1250 | 73 | |
| XM_007647805.1 | XP_007645995.1 | 7 - 8 | 1147 | 73 | ||
More Information
Set Membership: |
|

Tmlhe
Genomic Map