Assay Details
| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644413.1 | XP_007642603.1 | 1 - 2 | 175 | 90 | |
| GenBank mRNA | JI881166.1 | - | 1 - 2 | 116 | 90 | |
More Information
Set Membership: |
|

Nfkbib
Genomic Map