Assay Details
| Gene Symbol: Hras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509750.2 | XP_003509798.1 | 2 - 3 | 360 | 69 | |
| XM_007650381.1 | XP_007648571.1 | 2 - 3 | 363 | 69 | ||
| XM_007650382.1 | XP_007648572.1 | 2 - 3 | 365 | 69 | ||
| GenBank mRNA | JI869613.1 | - | 2 - 3 | 306 | 69 | |
| JP055473.1 | - | 2 - 3 | 306 | 69 | ||
More Information
Set Membership: |
|

Hras
Genomic Map