Assay Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511702.2 | XP_003511750.1 | 2 - 3 | 371 | 59 | |
| GenBank mRNA | JI866784.1 | - | 2 - 3 | 377 | 59 | |
| JP041302.1 | - | 2 - 3 | 377 | 59 | ||
More Information
Set Membership: |
|

Med27
Genomic Map