Assay Details
| Gene Symbol: LOC100768383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647575.1 | XP_007645765.1 | 4 - 5 | 302 | 87 | |
| GenBank mRNA | JI886640.1 | - | 1 - 2 | 82 | 87 | |
| JP064963.1 | - | 1 - 2 | 82 | 87 | ||
More Information
Set Membership: |
|

LOC100768383
Genomic Map