Assay Details
| Gene Symbol: LOC100768383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647575.1 | XP_007645765.1 | 6 - 7 | 734 | 97 | |
| GenBank mRNA | JI886641.1 | - | 1 - 2 | 304 | 97 | |
| JP064962.1 | - | 1 - 2 | 309 | 97 | ||
More Information
Set Membership: |
|

LOC100768383
Genomic Map