Assay Details
| Gene Symbol: LOC100768383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647575.1 | XP_007645765.1 | 7 - 8 | 839 | 85 | |
| GenBank mRNA | JI886641.1 | - | 2 - 3 | 409 | 85 | |
| JP064962.1 | - | 2 - 3 | 414 | 85 | ||
More Information
Set Membership: |
|

LOC100768383
Genomic Map