Assay Details
| Gene Symbol: Otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645197.1 | XP_007643387.1 | 3 - 4 | 385 | 86 | |
| XM_007645198.1 | XP_007643388.1 | 3 - 4 | 474 | 86 | ||
| XM_007645199.1 | XP_007643389.1 | 3 - 4 | 385 | 86 | ||
| GenBank mRNA | JI882907.1 | - | 3 - 4 | 349 | 86 | |
More Information
Set Membership: |
|

Otud6b
Genomic Map