Assay Details
| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646806.1 | XP_007644996.1 | 3 - 4 | 217 | 74 | |
| XM_007646807.1 | XP_007644997.1 | 3 - 4 | 193 | 74 | ||
| XM_007646808.1 | XP_007644998.1 | 3 - 4 | 199 | 74 | ||
| GenBank mRNA | JI888523.1 | - | 2 - 3 | 232 | 74 | |
More Information
Set Membership: |
|

Nudt3
Genomic Map