Assay Details
| Gene Symbol: Arglu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643859.1 | XP_007642049.1 | 3 - 4 | 400 | 116 | |
| XM_007643861.1 | XP_007642051.1 | 2 - 3 | 341 | 116 | ||
| GenBank mRNA | JP041763.1 | - | 1 - 2 | 90 | 116 | |
More Information
Set Membership: |
|

Arglu1
Genomic Map