Assay Details
| Gene Symbol: Rap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497137.2 | XP_003497185.1 | 6 - 7 | 638 | 78 | |
| GenBank mRNA | JI885426.1 | - | 6 - 7 | 676 | 78 | |
| JP059447.1 | - | 6 - 7 | 676 | 78 | ||
More Information
Set Membership: |
|

Rap1b
Genomic Map