Assay Details
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500314.2 | XP_003500362.1 | 4 - 5 | 732 | 75 | |
| XM_007641042.1 | XP_007639232.1 | 4 - 5 | 914 | 75 | ||
| XM_007641043.1 | XP_007639233.1 | 5 - 6 | 1055 | 75 | ||
| XM_007641044.1 | XP_007639234.1 | 4 - 5 | 913 | 75 | ||
| GenBank mRNA | JP043669.1 | - | 3 - 4 | 577 | 75 | |
More Information
Set Membership: |
|

Amz2
Genomic Map