Assay Details
| Gene Symbol: Tmed4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504757.2 | XP_003504805.1 | 3 - 4 | 424 | 66 | |
| GenBank mRNA | JI884060.1 | - | 3 - 4 | 423 | 66 | |
| JP060526.1 | - | 3 - 4 | 423 | 66 | ||
More Information
Set Membership: |
|

Tmed4
Genomic Map