Assay Details
| Gene Symbol: Prrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510039.1 | XP_003510087.1 | 8 - 9 | 1272 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrg3| Gene Symbol: Prrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510039.1 | XP_003510087.1 | 8 - 9 | 1272 | 114 | |
Set Membership: |
|