Assay Details
| Gene Symbol: LOC100758409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI884852.1 | - | 1 - 2 | 97 | 100 | |
| JP062511.1 | - | 1 - 2 | 97 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100758409| Gene Symbol: LOC100758409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI884852.1 | - | 1 - 2 | 97 | 100 | |
| JP062511.1 | - | 1 - 2 | 97 | 100 | ||