Assay Details
| Gene Symbol: Mrps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498194.2 | XP_003498242.1 | 1 - 2 | 91 | 60 | |
| GenBank mRNA | JI887162.1 | - | 1 - 2 | 98 | 60 | |
| JP051658.1 | - | 1 - 2 | 98 | 60 | ||
More Information
Set Membership: |
|

Mrps23
Genomic Map