Assay Details
| Gene Symbol: Tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007634464.1 | XP_007632654.1 | 5 - 6 | 267 | 53 | |
| XM_007634468.1 | XP_007632658.1 | 5 - 6 | 240 | 53 | ||
| XM_007634477.1 | XP_007632667.1 | 2 - 3 | 255 | 53 | ||
| XM_007634480.1 | XP_007632670.1 | 4 - 5 | 177 | 53 | ||
| XM_007634485.1 | XP_007632675.1 | 6 - 7 | 253 | 53 | ||
| XM_007634493.1 | XP_007632683.1 | 4 - 5 | 166 | 53 | ||
| XM_007634502.1 | XP_007632692.1 | 4 - 5 | 133 | 53 | ||
| XM_007634509.1 | XP_007632699.1 | 4 - 5 | 145 | 53 | ||
| XM_007634516.1 | XP_007632706.1 | 4 - 5 | 138 | 53 | ||
More Information
Set Membership: |
|

Tnni1
Genomic Map