Assay Details
| Gene Symbol: Cidea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645435.1 | XP_007643625.1 | 2 - 3 | 262 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cidea| Gene Symbol: Cidea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645435.1 | XP_007643625.1 | 2 - 3 | 262 | 62 | |
Set Membership: |
|