Assay Details
| Gene Symbol: Dag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500350.2 | XP_003500398.1 | 2 - 3 | 450 | 97 | |
| XM_007641129.1 | XP_007639319.1 | 2 - 3 | 460 | 97 | ||
| GenBank mRNA | JI885471.1 | - | 1 - 2 | 317 | 97 | |
More Information
Set Membership: |
|

Dag1
Genomic Map