Assay Details
| Gene Symbol: Znf652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503944.2 | XP_003503992.1 | 2 - 3 | 1407 | 80 | |
| GenBank mRNA | JI886191.1 | - | 1 - 2 | 34 | 80 | |
| JP048343.1 | - | 1 - 2 | 34 | 80 | ||
More Information
Set Membership: |
|

Znf652
Genomic Map